Learn More
Medchemexpress LLC Cddo-2p-im | 1883650-96-1 | MFCD03970221 | 98.1% | 618.81 g/mol | C39H46N4O3 | 5 MG
SDP
Catalog No. 5000263069
Encompass_Preferred
CDDO-2P-Im is an oleanane-derived synthetic triterpenoid research compound reported to have chemopreventive activity; studies show it can reduce the size and severity of lung tumors in mouse models. The compound is provided for research use and is typically used in vitro or in vivo with DMSO formulations. Characterization includes CAS 1883650-96-1, formula C39H46N4O3, and molecular weight 618.81 g/mol.
  • Potent chemopreventive activity demonstrated in murine cancer models.
  • High purity suitable for in vitro and in vivo studies.
  • Available as solid or pre-formulated solution in DMSO for convenience.
  • Characterized by CAS, formula, and molecular weight for traceability.
  • Supplied in small research-scale pack sizes for experimental workflows.
Catalog No. 50-002-63069 Supplier Medchemexpress LLC Supplier No. HY1263795MG
CDDO-2P-Im is an oleanane-derived synthetic triterpenoid research compound reported to have chemopreventive activity; studies show it can reduce the size and severity of lung tumors in mouse models. The compound is provided for research use and is typically used in vitro or in vivo with DMSO formulations. Characterization includes CAS 1883650-96-1, formula C39H46N4O3, and molecular weight 618.81 g/mol.
  • Potent chemopreventive activity demonstrated in murine cancer models.
  • High purity suitable for in vitro and in vivo studies.
  • Available as solid or pre-formulated solution in DMSO for convenience.
  • Characterized by CAS, formula, and molecular weight for traceability.
  • Supplied in small research-scale pack sizes for experimental workflows.