Learn More
Medchemexpress LLC CDDO-dhTFEA | 1191265-33-4 | 99.64% | 574.72 | 50 MG
SDP
Catalog No. 5000438695
Encompass_Preferred
CDDO-dhTFEA (RTA dh404) is a synthetic oleanane triterpenoid compound that potently activates Nrf2 and inhibits the pro-inflammatory transcription factor NF-κB. It restores hypertension (MAP), increases Nrf2 and the expression of its target genes, attenuates activation of NF-κB and transforming growth factor-β pathways, and reduces glomerulosclerosis, interstitial fibrosis, and inflammation in chronic kidney disease (CKD) rats. For research use only, not for sale to patients.
  • Potently activates Nrf2.
  • Inhibits the pro-inflammatory transcription factor NF-κB.
  • Attenuates activation of NF-κB and transforming growth factor-β pathways.
  • Reduces glomerulosclerosis, interstitial fibrosis, and inflammation in chronic kidney disease (CKD) rats.
  • Upregulates expression of antioxidant enzymes.
  • Protects pancreatic tissue and exhibits anti-inflammatory activity.
Catalog No. 50-004-38695 Supplier Medchemexpress LLC Supplier No. HY11267150MG
Add to Cart
Add to Cart
CDDO-dhTFEA (RTA dh404) is a synthetic oleanane triterpenoid compound that potently activates Nrf2 and inhibits the pro-inflammatory transcription factor NF-κB. It restores hypertension (MAP), increases Nrf2 and the expression of its target genes, attenuates activation of NF-κB and transforming growth factor-β pathways, and reduces glomerulosclerosis, interstitial fibrosis, and inflammation in chronic kidney disease (CKD) rats. For research use only, not for sale to patients.
  • Potently activates Nrf2.
  • Inhibits the pro-inflammatory transcription factor NF-κB.
  • Attenuates activation of NF-κB and transforming growth factor-β pathways.
  • Reduces glomerulosclerosis, interstitial fibrosis, and inflammation in chronic kidney disease (CKD) rats.
  • Upregulates expression of antioxidant enzymes.
  • Protects pancreatic tissue and exhibits anti-inflammatory activity.