Learn More
Medchemexpress LLC HY-112671 5mg Medchemexpress, CDDO-dhTFEA CAS:1191265-33-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1126715MG
Encompass_Preferred
Medchemexpress, HY-112671 5mg CDDO-dhTFEA CAS:1191265-33-4 CDDO-dhTFEA (RTA dh404) is a synthetic oleanane triterpenoid compound which potently activates Nrf2 and inhibits the pro-inflammatory transcription factor NF-κB. CDDO-dhTFEA restores hypertension (MAP), increases Nrf2 and expression of its target genes, attenuates activation of NF-κB and transforming growth factor-β pathways, and reduces glomerulosclerosis, interstitial fibrosis and inflammation in the chronic kidney disease (CKD) rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9136