Learn More
Medchemexpress LLC HY-12213 5mg Medchemexpress, CDDO-EA CAS:932730-51-3 Purity:>98%
SDP
Catalog No. 502080980
Encompass_Preferred
Medchemexpress, HY-12213 5mg CDDO-EA CAS:932730-51-3 CDDO-EA is an NF-E2 related factor 2/antioxidant response element (Nrf2/ARE) activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0980 Supplier Medchemexpress LLC Supplier No. HY122135MG
Medchemexpress, HY-12213 5mg CDDO-EA CAS:932730-51-3 CDDO-EA is an NF-E2 related factor 2/antioxidant response element (Nrf2/ARE) activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.