Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cddo-ea | 932730-51-3 | 99.8% | 1 ML
SDP

Catalog No. 5000232535
Encompass_Preferred

CDDO-EA (CDDO ethyl amide) is a synthetic triterpenoid that activates the NF-E2-related factor 2/antioxidant response element (Nrf2/ARE) signaling pathway. It is used in research to modulate oxidative stress, inflammation, and apoptosis, and is supplied as a solid or as a 10 mM solution in DMSO for cell-based assays.

  • Activates Nrf2/ARE signaling and modulates oxidative stress.
  • Available as a solid or a 10 mM solution in DMSO.
  • High purity suitable for research use.
  • Molecular weight 518.73 g/mol; formula C33H46N2O3.
  • Commonly used in in vitro studies of inflammation and apoptosis.

Catalog No. 50-002-32535 Supplier Medchemexpress LLC Supplier No. HY1221310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CDDO-EA (CDDO ethyl amide) is a synthetic triterpenoid that activates the NF-E2-related factor 2/antioxidant response element (Nrf2/ARE) signaling pathway. It is used in research to modulate oxidative stress, inflammation, and apoptosis, and is supplied as a solid or as a 10 mM solution in DMSO for cell-based assays.

  • Activates Nrf2/ARE signaling and modulates oxidative stress.
  • Available as a solid or a 10 mM solution in DMSO.
  • High purity suitable for research use.
  • Molecular weight 518.73 g/mol; formula C33H46N2O3.
  • Commonly used in in vitro studies of inflammation and apoptosis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.