Learn More
Medchemexpress LLC HY-15725 10mg , CDDO-Im RTA-403;TP-235;CDDO-Imidazolide CAS:443104-02-7 Purity:98%
SDP
Catalog No. 501494044
Encompass_Preferred
Medchemexpress, HY-15725 10mg CDDO-Im RTA-403;TP-235;CDDO-Imidazolide CAS:443104-02-7 Formula:C34H43N3O3 Ki: 232 nM (PPARα), 344 nM (PPARγ) Nrf2 Purity:98% CDDO-Im (CDDO-imidazolide) is an activator of Nrf2 and PPAR , with K i s of 232 and 344 nM for PPARα and PPARγ, also has anti-cancer and anti-inflammatory activities. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4044 Supplier Medchemexpress LLC Supplier No. HY1572510MG
Medchemexpress, HY-15725 10mg CDDO-Im RTA-403;TP-235;CDDO-Imidazolide CAS:443104-02-7 Formula:C34H43N3O3 Ki: 232 nM (PPARα), 344 nM (PPARγ) Nrf2 Purity:98% CDDO-Im (CDDO-imidazolide) is an activator of Nrf2 and PPAR , with K i s of 232 and 344 nM for PPARα and PPARγ, also has anti-cancer and anti-inflammatory activities. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.