Learn More
Eurofins Discoverx CDDO Methyl Ester, 5 mg. (92-1147)
Supplier:  Eurofins Discoverx 921147
Encompass
CDDO Methyl Ester (bardoxolone methyl) is a semi-synthetic triterpenoid developed from the oleanolic acid scaffold that induces apoptosis and differentiation in tumor cells. Agonist of the KEAP1-Nrf2 pathway, thus inducing expression of antioxidant proteins and protection against oxidative damage from injury and inflammation. Also an antagonist of the NF-kB pro-inflammatory transcription factor and PPARG. CDDO methyl ester underwent clinical trials for type 2 diabetes treatment, due to its potent antidiabetic activity, but was confirmed to enhance cardiac disease during a phase 2 trial in October 2012. Provided as 5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9678