Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 999MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: 4A2. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: LS174T, Raji, HT29, SK-BR3 cells. Prostate or Colon carcinomas. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length E-Cadherin protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 120-80kDa (Mature) ;135kDa (Precursor).Chromosome: 16q22.1. Gene Symbol: CDH1. Gene ID: 999. Ensemble: ENSG00000039068. Uniprot: NP_001304114.
Catalog No. 50-188-5400