Learn More
enQuire Bioreagents Ksp-Cadherin Recombinant Monoclonal [Clone CDH16/1532R], Size: 100ug
Supplier:  enQuire Bioreagents 1014RBM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: CDH16/1532R. Clonality: Monoclonal. Host: Rabbit. Isotype: IgG kappa. Positive Control: Normal kidney or renal cell carcinoma. Cellular Staining: Cell Surface with some cytoplasmic. Buffer: 10mM PBS with or without 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length CDH16 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 130kDa.Chromosome: 16q22.1. Gene Symbol: CDH16. Gene ID: 1014. Ensemble: ENSG00000166589. Uniprot: NP_001191674.
Catalog No. 50-188-4554