Learn More
Medchemexpress LLC CDH16 Human HEK293 50ug
SDP
Catalog No. 5000180076
Encompass_Preferred
CDH16 a calcium-dependent cell adhesion protein belongs to the cadherin family It exhibits homophilic interactions potentially contributing to cell-type sorting and regulating cell-cell adhesions for cellular organization CDH16 Protein Human (HEK293 His) is the recombinant human-derived CDH16 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80076 Supplier Medchemexpress LLC Supplier No. HYP7350750UG
CDH16 a calcium-dependent cell adhesion protein belongs to the cadherin family It exhibits homophilic interactions potentially contributing to cell-type sorting and regulating cell-cell adhesions for cellular organization CDH16 Protein Human (HEK293 His) is the recombinant human-derived CDH16 protein expressed by HEK293 with C-His labeled tag