Learn More
Abnova™ CDH17 Split FISH Probe
Catalog No. 89999067 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 uL
Catalog No. Quantity
89-999-067 100 uL
1 options
Catalog No. 89-999-067 Supplier Abnova™ Supplier No. FS0070
Add to Cart
Add to Cart
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Specifications
Quantity 100 uL
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
For Use With (Application) FISH-Ce
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe