Learn More
Medchemexpress LLC Cadherin-19 (human) Flag-MBP fusion protein | >95.0% | 20UG
SDP
Catalog No. 5000175374
Encompass_Preferred
Recombinant human cadherin-19 extracellular fragment supplied as an N-terminal Flag-MBP fusion and provided as a lyophilized powder for cell adhesion and binding studies, including cell sorting and biochemical assays. The product includes formulation and storage guidance to help preserve activity and minimize endotoxin.
Catalog No. 50-001-75374 Supplier Medchemexpress LLC Supplier No. HYP70097320UG
Recombinant human cadherin-19 extracellular fragment supplied as an N-terminal Flag-MBP fusion and provided as a lyophilized powder for cell adhesion and binding studies, including cell sorting and biochemical assays. The product includes formulation and storage guidance to help preserve activity and minimize endotoxin.