Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cdh19 protein, human (Flag-MBP) | >95.0% | 50UG
SDP

Catalog No. 5000176205
Encompass_Preferred

Recombinant human CDH19 protein (N-terminal MBP and Flag tagged), corresponding to residues G44-H569, supplied as a lyophilized powder for research use. Expressed in E. coli, this partial cadherin-19 construct is provided at 50 μg with >95% purity and an approximate molecular weight of 100.89 kDa. The product is lyophilized from buffered solution with trehalose; storage and reconstitution guidance are provided by the manufacturer.

  • Recombinant fragment corresponding to residues G44-H569
  • N-terminal MBP and Flag tags to aid purification and detection
  • Expressed in E. coli and supplied as lyophilized powder
  • Purity greater than 95% as determined by reducing SDS-PAGE
  • Molecular weight approximately 100.89 kDa
  • Formulated with trehalose as lyoprotectant in buffered solution
  • Store at -20°C; do not reconstitute below 100 μg/mL in ddH2O

Catalog No. 50-001-76205 Supplier Medchemexpress LLC Supplier No. HYP70097350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human CDH19 protein (N-terminal MBP and Flag tagged), corresponding to residues G44-H569, supplied as a lyophilized powder for research use. Expressed in E. coli, this partial cadherin-19 construct is provided at 50 μg with >95% purity and an approximate molecular weight of 100.89 kDa. The product is lyophilized from buffered solution with trehalose; storage and reconstitution guidance are provided by the manufacturer.

  • Recombinant fragment corresponding to residues G44-H569
  • N-terminal MBP and Flag tags to aid purification and detection
  • Expressed in E. coli and supplied as lyophilized powder
  • Purity greater than 95% as determined by reducing SDS-PAGE
  • Molecular weight approximately 100.89 kDa
  • Formulated with trehalose as lyoprotectant in buffered solution
  • Store at -20°C; do not reconstitute below 100 μg/mL in ddH2O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.