Learn More
Aviva Systems Biology Corp CDH22 Antibody - N-terminal region (ARP49648_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49648_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDH22. It was validated on Western Blot. . Alternate names: C20orf25, MGC39564, dJ998H6.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-441