Learn More
Aviva Systems Biology Corp CDH4 Antibody - N-terminal region (ARP45172_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45172_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDH4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CDH4. Alternate names: CAD4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCADApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53993