Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cdi

Catalog No. 11553325KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
25 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-553-325KG 25 kg
11-553-3500G 500 g
11-553-31KG 1 kg
11-553-3100G 100 g
11-553-35G 5 g
11-553-325G 25 g
11-553-310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 11-553-325KG Supplier Sigma Aldrich Supplier No. 11553325KG
Add to Cart
Add to Cart
Request a Quote

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cdi

Specifications

CAS 530-62-1
Melting Point 117°C to 122°C
Molecular Formula C7H6N4O
Linear Formula C7H6N4O
MDL Number MFCD00005286
Quantity 25 kg
Synonym 1,1'-Carbonyldiimidazole
Molecular Weight (g/mol) 162.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.