Learn More
Bioworld CDI-ACTIVATED SEPAROPOREA
Supplier:  Bioworld 201810845
Encompass
CDI-ACTIVATED SEPAROPORE (AGAROSE) 6B: CDI-activated Separopore6B Pre-activated medium for immobilization of ligands containing primary amines.; N,N\'-Carbonyl diimidazole (CDI) mediated activation of Separopore matrix to allow coupling...
Catalog No. 50-197-887