Learn More
Enzo Life Sciences Cdk/Crk inhibitor (1mg). CAS: 784211-09-2
SDP
Catalog No. 502013423
Encompass_Preferred
Potent, selective, and ATP-competitive inhibitor of Cdks. The Cdk/Crk inhibitor is a broad-spectrum growth inhibitor toward tumor cells. Alternative name: 1-(2,6-Dichlorophenyl)-1,5-dihydro-6-((4-(2-hydroxyethoxy)phenyl)methyl)-3-(1-methylethyl)-4H-pyrazolo[3,4-d]pyrimidin-4-one, RGB-286147, Cdk7 Inhibitor IV. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3423 Supplier Enzo Life Sciences Supplier No. ENZCHM1310001
Potent, selective, and ATP-competitive inhibitor of Cdks. The Cdk/Crk inhibitor is a broad-spectrum growth inhibitor toward tumor cells. Alternative name: 1-(2,6-Dichlorophenyl)-1,5-dihydro-6-((4-(2-hydroxyethoxy)phenyl)methyl)-3-(1-methylethyl)-4H-pyrazolo[3,4-d]pyrimidin-4-one, RGB-286147, Cdk7 Inhibitor IV. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.