Learn More
Medchemexpress LLC HY-126534A 5mg Medchemexpress, CDK ligand for PROTAC (hydrochloride) CAS: Purity:>98%
SDP
Catalog No. 501969733
Encompass_Preferred
Medchemexpress, HY-126534A 5mg CDK ligand for PROTAC (HCl) CAS: CDK ligand for PROTAC hydrochloride is a CDK inhibitor with antitumor activity. CDK ligand for PROTAC hydrochloride and a CRBN ligand have been used to design PROTAC CDK4/6 degrader. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9733 Supplier Medchemexpress LLC Supplier No. HY126534A5MG
Medchemexpress, HY-126534A 5mg CDK ligand for PROTAC (HCl) CAS: CDK ligand for PROTAC hydrochloride is a CDK inhibitor with antitumor activity. CDK ligand for PROTAC hydrochloride and a CRBN ligand have been used to design PROTAC CDK4/6 degrader. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.