Learn More
TARGETMOL CHEMICALS INC CDK1/2 INHIBITOR III 5MG
SDP
Catalog No. 502581272
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cdk1/2 Inhibitor III is a selective Cdk1/2 inhibitor with an IC50 value of 2.1 uM against CDK1/cyclin B. purity: 99%
Catalog No. 50-258-1272 Supplier TARGETMOL CHEMICALS INC Supplier No. T149145MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cdk1/2 Inhibitor III is a selective Cdk1/2 inhibitor with an IC50 value of 2.1 uM against CDK1/cyclin B. purity: 99%