Learn More
ABclonal Technology CDK1/2 Rabbit PolymAb
SDP
Catalog No. 5000012899
Encompass_Preferred
This gene encodes a member of a family of serine threonine protein kinases that participate in cell cycle regulation The encoded protein is the catalytic subunit of the cyclin-dependent protein kinase complex which regulates progression through the cell cycle Activity of this protein is especially critical during the G1 to S phase transition This protein associates with and regulated by other subunits of the complex including cyclin A or E CDK inhibitor p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B) Alternative splicing results in multiple transcript variants
Catalog No. 50-000-12899 Supplier ABclonal Technology Supplier No. A26696PM20UL
This gene encodes a member of a family of serine threonine protein kinases that participate in cell cycle regulation The encoded protein is the catalytic subunit of the cyclin-dependent protein kinase complex which regulates progression through the cell cycle Activity of this protein is especially critical during the G1 to S phase transition This protein associates with and regulated by other subunits of the complex including cyclin A or E CDK inhibitor p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B) Alternative splicing results in multiple transcript variants