Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 983MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: POH-1; same as cdc2.1. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: HeLa or MCF-7 Cells or Tonsil. Cellular Staining: Nuclear, Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Purified recombinant human p34cdc2 fusion protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 34kDa.Chromosome: 10q21.2. Gene Symbol: CDC2. Gene ID: 983. Ensemble: ENSG00000170312. Uniprot: NP_001163878.
Catalog No. 50-188-5390