Learn More
Medchemexpress LLC Cdk1/Cyc B-IN-1 | 2459916-56-2 | 98.5% | 353.85 | 5 MG
SDP
Catalog No. 5000360694
Encompass_Preferred
CDK1/Cyc B-IN-1 is a selective inhibitor of the CDK1/Cyc B complex, exhibiting an IC50 of 97 nM. This compound induces apoptosis and G2/M cell cycle arrest. It demonstrates broad-spectrum cytotoxic activity across various cancer cell lines.
  • Selective CDK1/Cyc B complex inhibitor.
  • IC50 of 97 nM against CDK1-Cyclin B.
  • Triggers apoptosis.
  • Induces G2/M cell cycle arrest.
  • Exhibits broad-spectrum cytotoxic action against cancer cell lines.
  • Shows cytotoxic activity against HCT-116 and WI-38 cells with IC50 values of 5.33 ± 0.69 μM and 21.69 ± 1.04 μM respectively.
Catalog No. 50-003-60694 Supplier Medchemexpress LLC Supplier No. HY1476465MG
CDK1/Cyc B-IN-1 is a selective inhibitor of the CDK1/Cyc B complex, exhibiting an IC50 of 97 nM. This compound induces apoptosis and G2/M cell cycle arrest. It demonstrates broad-spectrum cytotoxic activity across various cancer cell lines.
  • Selective CDK1/Cyc B complex inhibitor.
  • IC50 of 97 nM against CDK1-Cyclin B.
  • Triggers apoptosis.
  • Induces G2/M cell cycle arrest.
  • Exhibits broad-spectrum cytotoxic action against cancer cell lines.
  • Shows cytotoxic activity against HCT-116 and WI-38 cells with IC50 values of 5.33 ± 0.69 μM and 21.69 ± 1.04 μM respectively.