Learn More
Medchemexpress LLC Cdk1/Cyc B-IN-1 | 2459916-56-2 | 98.5% | 353.85 | 50 MG
SDP
Catalog No. 5000362219
Encompass_Preferred
CDK1/Cyc B-IN-1 (Compound 5) is a selective CDK1/Cyc B complex inhibitor with an IC50 of 97 nM. It triggers apoptosis and G2/M cell cycle arrest and demonstrates broad-spectrum cytotoxic action against cancer cell lines. In vitro, it is soluble in DMSO at 100 mg/mL.
  • Induces apoptosis and G2/M cell cycle arrest
  • Exhibits cytotoxic activity against HCT-116 cells (IC50 of 5.33 ± 0.69 μM)
  • Exhibits cytotoxic activity against WI-38 cells (IC50 of 21.69 ± 1.04 μM)
  • Shows antiproliferative activity against various human cancer cell lines
Catalog No. 50-003-62219 Supplier Medchemexpress LLC Supplier No. HY14764650MG
CDK1/Cyc B-IN-1 (Compound 5) is a selective CDK1/Cyc B complex inhibitor with an IC50 of 97 nM. It triggers apoptosis and G2/M cell cycle arrest and demonstrates broad-spectrum cytotoxic action against cancer cell lines. In vitro, it is soluble in DMSO at 100 mg/mL.
  • Induces apoptosis and G2/M cell cycle arrest
  • Exhibits cytotoxic activity against HCT-116 cells (IC50 of 5.33 ± 0.69 μM)
  • Exhibits cytotoxic activity against WI-38 cells (IC50 of 21.69 ± 1.04 μM)
  • Shows antiproliferative activity against various human cancer cell lines