Learn More
Cayman Chemical Cdk1 InhIbItr 1mg
SDP
Catalog No. 502986244
Encompass_Preferred
An inhibitor of Cdk1 (IC50 = 5.8 µM); selective for Cdk1 over Cdk5 and GSK3β (IC50s = 25 and >100 µM, respectively); inhibits HeLa cell proliferation (IC50 = 2 µM)
Catalog No. 50-298-6244 Supplier Cayman Chemical Supplier No. 215011 MG
An inhibitor of Cdk1 (IC50 = 5.8 µM); selective for Cdk1 over Cdk5 and GSK3β (IC50s = 25 and >100 µM, respectively); inhibits HeLa cell proliferation (IC50 = 2 µM)