Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CDK11 (Inactive), GST Tag Recombinant Protein

Catalog No. PV6283
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6283 10 μg
PV6284 100 μg
PV6285 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV6283 Supplier Invitrogen™ Supplier No. PV6283
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human CDK11 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for CDK11 (Inactive) is NP_055891.1.

CDC2L6 is one of the components of the Mediator coactivator complex. The Mediator complex is a multiprotein complex required for transcriptional activation by DNA binding transcription factors of genes transcribed by RNA polymerase II. The protein encoded by this gene is similar to cyclin-dependent kinase 8 which can also be a component of the Mediator complex.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BWU1
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 23097
Molecular Weight (g/mol) 84.2 kDa
Name Human CDK11 (Inactive), GST Tag
Quantity 10 μg
Gene Alias 2700084L06Rik; AI845279; AW228747; bA346C16.3; Cdc2l6; CDC2-related protein kinase 6; Cdk11; Cdk19; CDK8-like cyclin-dependent kinase; cell division cycle 2-like 6 (CDK8-like); cell division cycle 2-like protein kinase 6; Cell division protein kinase 19; cyclin dependent kinase 19; cyclin dependent kinase 19 variant 2; cyclin-dependent kinase (CDC2-like) 11; cyclin-dependent kinase 11; Cyclin-dependent kinase 19; death-preventing kinase; KIAA1028; mKIAA1028
Common Name CDC2L6
Protein Length 1-795
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.