Learn More
Medchemexpress LLC CDK12-IN-2 | 2244987-03-7 | 99.4% | 532.64 | 10 MG
SDP
Catalog No. 5000436395
Encompass_Preferred
CDK12-IN-2 is a potent, selective, and nanomolar CDK12 inhibitor with good physicochemical properties. It also acts as a strong CDK13 inhibitor because CDK13 is the closest homologue of CDK12. It demonstrates excellent kinase selectivity for CDK12 over CDK2, 9, 8, and 7. This compound inhibits the phosphorylation of Ser2 in the C-terminal domain of RNA polymerase II and can be used as an excellent chemical probe for functional studies of CDK12.
  • Potent, selective, and nanomolar CDK12 inhibitor
  • Strong CDK13 inhibitor
  • Excellent kinase selectivity over CDK2, 9, 8, and 7
  • Inhibits the phosphorylation of Ser2 in the C-terminal domain of RNA polymerase II
  • Can be used as an excellent chemical probe for functional studies of CDK12
Catalog No. 50-004-36395 Supplier Medchemexpress LLC Supplier No. HY11262610MG
CDK12-IN-2 is a potent, selective, and nanomolar CDK12 inhibitor with good physicochemical properties. It also acts as a strong CDK13 inhibitor because CDK13 is the closest homologue of CDK12. It demonstrates excellent kinase selectivity for CDK12 over CDK2, 9, 8, and 7. This compound inhibits the phosphorylation of Ser2 in the C-terminal domain of RNA polymerase II and can be used as an excellent chemical probe for functional studies of CDK12.
  • Potent, selective, and nanomolar CDK12 inhibitor
  • Strong CDK13 inhibitor
  • Excellent kinase selectivity over CDK2, 9, 8, and 7
  • Inhibits the phosphorylation of Ser2 in the C-terminal domain of RNA polymerase II
  • Can be used as an excellent chemical probe for functional studies of CDK12