Learn More
Medchemexpress LLC HY-112261 10mg Medchemexpress, CDK12-IN-3 CAS:2220184-50-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11226110MG
Encompass_Preferred
Medchemexpress, HY-112261 10mg CDK12-IN-3 CAS:2220184-50-7 CDK12-IN-3 is a potent and selective CDK12 inhibitor with an IC50 of 491 nM in enzymatic assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9067