Learn More
Medchemexpress LLC CDK12-IN-3 | 2220184-50-7 | 99.9% | 470.52 | 50 MG
SDP
Catalog No. 5000437430
Encompass_Preferred
CDK12-IN-3 is a potent and selective CDK12 inhibitor with an IC50 of 491 nM in enzymatic assay. It is for research use only and not sold to patients.
  • Potent and selective CDK12 inhibitor.
  • IC50 of 491 nM in enzymatic assay.
  • Shows potent inhibition of phosphorylation of Ser2 on the CTD repeat domain of RNA Pol II (at 0.1 μM).
  • Exhibits growth inhibition of OV90 cells and acute cytotoxicity to THP1 cells.
Catalog No. 50-004-37430 Supplier Medchemexpress LLC Supplier No. HY11226150MG
CDK12-IN-3 is a potent and selective CDK12 inhibitor with an IC50 of 491 nM in enzymatic assay. It is for research use only and not sold to patients.
  • Potent and selective CDK12 inhibitor.
  • IC50 of 491 nM in enzymatic assay.
  • Shows potent inhibition of phosphorylation of Ser2 on the CTD repeat domain of RNA Pol II (at 0.1 μM).
  • Exhibits growth inhibition of OV90 cells and acute cytotoxicity to THP1 cells.