Learn More
Medchemexpress LLC Cdk12-in-3 | 2220184-50-7 | MFCD31746915 | 99.9% | 470.52 g/mol | C23H28F2N8O | 5 MG
SDP
Catalog No. 5000263146
Encompass_Preferred
CDK12-IN-3 is a small-molecule inhibitor selective for cyclin-dependent kinase 12 (CDK12) used in research on transcriptional regulation and DNA damage response. It inhibits CDK12 enzymatic activity (reported IC50 491 nM), impacts phosphorylation of the RNA polymerase II CTD Ser2, and is supplied in high purity for biochemical and cell-based studies.
Catalog No. 50-002-63146 Supplier Medchemexpress LLC Supplier No. HY1122615MG
CDK12-IN-3 is a small-molecule inhibitor selective for cyclin-dependent kinase 12 (CDK12) used in research on transcriptional regulation and DNA damage response. It inhibits CDK12 enzymatic activity (reported IC50 491 nM), impacts phosphorylation of the RNA polymerase II CTD Ser2, and is supplied in high purity for biochemical and cell-based studies.