Learn More
Medchemexpress LLC CDK12-IN-E9 100mg | 2020052-55-3 | 434.53 | C24H30N6O2 | 100 MG
SDP
Catalog No. 5000226787
Encompass_Preferred
CDK12-IN-E9 is a research-grade small-molecule inhibitor that covalently inhibits CDK12 and has non-covalent activity against CDK9. Supplied as a white to off-white solid powder, it is intended for biochemical and cellular studies of cell cycle and DNA damage pathways. Solubility and storage data support use in both in vitro and in vivo formulations.
Catalog No. 50-002-26787 Supplier Medchemexpress LLC Supplier No. HY117203A100MG
CDK12-IN-E9 is a research-grade small-molecule inhibitor that covalently inhibits CDK12 and has non-covalent activity against CDK9. Supplied as a white to off-white solid powder, it is intended for biochemical and cellular studies of cell cycle and DNA damage pathways. Solubility and storage data support use in both in vitro and in vivo formulations.