Learn More
Medchemexpress LLC CDK12-IN-E9 50mg | 2020052-55-3 | 434.53 | C24H30N6O2 | 50 MG
SDP
Catalog No. 5000228319
Encompass_Preferred
CDK12-IN-E9 is a selective covalent inhibitor of cyclin-dependent kinase 12 (CDK12) that also shows non-covalent inhibition of CDK9. Supplied as a research-grade small molecule, it is used in studies of cell-cycle regulation and the DNA damage response. Typical formats include a solid powder and a ready-to-use DMSO solution.
Catalog No. 50-002-28319 Supplier Medchemexpress LLC Supplier No. HY117203A50MG
CDK12-IN-E9 is a selective covalent inhibitor of cyclin-dependent kinase 12 (CDK12) that also shows non-covalent inhibition of CDK9. Supplied as a research-grade small molecule, it is used in studies of cell-cycle regulation and the DNA damage response. Typical formats include a solid powder and a ready-to-use DMSO solution.