Learn More
Aviva Systems Biology Corp Cdk14 Antibody - C-terminal region (ARP61958_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61958_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cdk14. It was validated on Western Blot. . Alternate names: Pftk1, mKIAA0834ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-520