Learn More
Cayman Chemical Cdk15 InhIbItr 5mg
SDP
Catalog No. 502979417
Encompass_Preferred
Inhibitor of Cdk1/cyclin B and Cdk/p25 (IC50s = 600 and 400 nM, respectively)
Catalog No. 50-297-9417 Supplier Cayman Chemical Supplier No. 187405 MG
Add to Cart
Add to Cart
Inhibitor of Cdk1/cyclin B and Cdk/p25 (IC50s = 600 and 400 nM, respectively)