Learn More
Sino Biological Recombinant Human CLK4 Protein (I363V, GST Tag), Active
Catalog No. 502575499
Encompass_Preferred
Recombinant human CLK4 (I363V) (70-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5499 Supplier Sino Biological Supplier No. C6112G50
Recombinant human CLK4 (I363V) (70-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.