Learn More
Carnabio Usa Inc CDK2/CYCA2/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 041035
Encompass
CDK2/CycA2, Full-length, Wild type, Amino Acid 1-298, NP_001789.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0363