Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CDK2/Cyclin A, His Tag Recombinant Protein

Catalog No. PR4448A
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR4448A 1 mg
PR4448B 100 μg
PV3267 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR4448A Supplier Invitrogen™ Supplier No. PR4448A
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.

CDK2/cyclinO is a member of the cyclin dependent protein kinase family complexed to CyclinO. CyclinO was used as a specific protein isoform name for the UNG gene which is localized exclusively in the nucleus. In addition, CyclinO was reported to shows significant uracil-DNA glycosylase activity. CDK2/cyclinO protein levels increase during G1 phase and the protein is turned over during the course of cell cycle. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P24941
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 20mM tris with 2mM DTT, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 20% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1017
Molecular Weight (g/mol) 91.3 kDa
Name Human CDK2/Cyclin A, His Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Source Human
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias A630093N05Rik; CDC28; CDC2A; cdc2-related protein kinase; CDK1; CDK2; CDKN2; Cell division protein kinase 2; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 2; EC 2.7.11.22; EC 2.7.11.23; kinase Cdc2; MPF; p33 protein kinase; p33(CDK2); p33CDK2; p34 protein kinase
Common Name CDK2
Gene Symbol CDK2
Kinase Family CDK and CHK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: CMCG Group
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Research Category Cell Cycle & Proliferation, Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.