Learn More
Eurofins Discoverx Cdk2/Cyclin E, active, 250 µg. (14-475M)
Supplier:  Eurofins Discoverx 14475M
Encompass
Complex of recombinant, human full length cdk2 containing a C-terminal 6xHis-tag, and recombinant, human full length cyclin E containing an N-terminal GST-tag. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8842