Learn More
Enzo Life Sciences CDK2 / Cyclin E (human), (recombinant) (GST-tag) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE2690010
Encompass_Preferred
Active serine/threonine kinase, using histone H1 as substrate. Active serine/threonine kinase, using histone H1 as substrate. Alternative name: Cyclin dependent kinase 2 / Cyclin B. Formulation: Liquid. In 50mM TRIS-HCl, pH 8.0, containing 100mM sodium chloridel, 5mM DTT, 15mM reduced glutathione and 20% glycerol. Source: Produced in insect cells. GST fusion proteins containing CDK2 (aa 1-298) and residues cyclin E (aa 1-394). Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-1964