Learn More
AURORA BIOLABS LLC Aurora Biolabs, Kinase Assay Kit, CDK2, CyclinA2, 384 Reactions
Catalog No. NC3915273
Encompass_Preferred
The CDK2/CyclinA2 Kinase Assay Kit is designed for the quantitative analysis of CDK2/CyclinA2 kinase activity. This bioluminescent assay relies on the measurement of ATP consumption to determine kinase activity, utilizing a coupled luciferase reaction system.
  • Provides sufficient reagents for 384 reactions
  • Includes recombinant human CDK2/CyclinA2 enzyme
  • Contains 5 mM substrate peptide and 1 mM ATP
  • Supplied with D-luciferin and firefly luciferase for detection
  • Includes 384-well microplate
  • Optimized for use with a microplate reader capable of measuring luminescence
  • Requires an adjustable micro-pipettor and sterile tips
  • Intended for research use only
Catalog No. NC3915273 Supplier AURORA BIOLABS LLC Supplier No. C352A2KA
The CDK2/CyclinA2 Kinase Assay Kit is designed for the quantitative analysis of CDK2/CyclinA2 kinase activity. This bioluminescent assay relies on the measurement of ATP consumption to determine kinase activity, utilizing a coupled luciferase reaction system.
  • Provides sufficient reagents for 384 reactions
  • Includes recombinant human CDK2/CyclinA2 enzyme
  • Contains 5 mM substrate peptide and 1 mM ATP
  • Supplied with D-luciferin and firefly luciferase for detection
  • Includes 384-well microplate
  • Optimized for use with a microplate reader capable of measuring luminescence
  • Requires an adjustable micro-pipettor and sterile tips
  • Intended for research use only