Learn More
AURORA BIOLABS LLC CDK2/CyclinE1 Kinase Assay Kit,384 rxn
Catalog No. NC4007628
Encompass_Preferred
The CDK2/CyclinE1 assay kit is a luminescence based assay by measuring remaining ATP amount after the kinase reaction using firefly luciferase. The assay kit is designed in 384-well format and is convenient for high-throughput compound screening. CDK2 uses ATP to phosphorylate its substrate peptide to produce the phosphorylated peptide and ADP. Its kinase activity then can be estimated by measuring the amount of the remaining ATP.
Catalog No. NC4007628 Supplier AURORA BIOLABS LLC Supplier No. C352E1KA384
The CDK2/CyclinE1 assay kit is a luminescence based assay by measuring remaining ATP amount after the kinase reaction using firefly luciferase. The assay kit is designed in 384-well format and is convenient for high-throughput compound screening. CDK2 uses ATP to phosphorylate its substrate peptide to produce the phosphorylated peptide and ADP. Its kinase activity then can be estimated by measuring the amount of the remaining ATP.