Learn More
Medchemexpress LLC CDK2 degrader 5 | 2923574-11-0 | 99.2% | 497.54 | 25 MG
SDP
Catalog No. 5000394058
Encompass_Preferred
CDK2 degrader 5 (compound 12) is a degrader for CDK2, with a Dmax of >50% and ≤80% in HiBiT Assay. It plays an important role in cancer research.
  • Degrader for CDK2.
  • Dmax of >50% and ≤80% in HiBiT Assay.
  • Used for research purposes only.
  • Solid, white to off-white appearance.
  • Plays an important role in cancer research.
Catalog No. 50-003-94058 Supplier Medchemexpress LLC Supplier No. 25MGHY169912
CDK2 degrader 5 (compound 12) is a degrader for CDK2, with a Dmax of >50% and ≤80% in HiBiT Assay. It plays an important role in cancer research.
  • Degrader for CDK2.
  • Dmax of >50% and ≤80% in HiBiT Assay.
  • Used for research purposes only.
  • Solid, white to off-white appearance.
  • Plays an important role in cancer research.