Learn More
Medchemexpress LLC CDK2-IN-4 | 1316652-41-1 | 99.3% | 326.37 g/mol | C16H14N4O2S | 5 MG
SDP
Catalog No. 5000227031
Encompass_Preferred
NCC-149 is a selective histone deacetylase 8 (HDAC8) inhibitor used in cell biology research, including studies of neural differentiation. The product is provided with supporting documentation for purity, handling, and storage, and is available both as a solid and as a DMSO solution.
  • Selective HDAC8 inhibitor for neural differentiation studies.
  • Documented purity approximately 99.3% based on certificate of analysis.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Datasheet, certificate of analysis, and safety data sheet provided for quality and safety information.
  • Storage recommendations included for powdered material and solutions.
Catalog No. 50-002-27031 Supplier Medchemexpress LLC Supplier No. HY1173485MG
NCC-149 is a selective histone deacetylase 8 (HDAC8) inhibitor used in cell biology research, including studies of neural differentiation. The product is provided with supporting documentation for purity, handling, and storage, and is available both as a solid and as a DMSO solution.
  • Selective HDAC8 inhibitor for neural differentiation studies.
  • Documented purity approximately 99.3% based on certificate of analysis.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Datasheet, certificate of analysis, and safety data sheet provided for quality and safety information.
  • Storage recommendations included for powdered material and solutions.