Learn More
TARGETMOL CHEMICALS INC CDK2 INHIBITOR II 5MG
SDP
Catalog No. 502580159
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cdk2 Inhibitor II is a selective and potent CDK2 inhibitor50 at 60 nM. purity: 98%
Catalog No. 50-258-0159 Supplier TARGETMOL CHEMICALS INC Supplier No. T369335MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cdk2 Inhibitor II is a selective and potent CDK2 inhibitor50 at 60 nM. purity: 98%