Learn More
Cayman Chemical CDK2 Inhibitor 73, Purity: ≥98%, CAS: 2079895-42-2, 5 mg
SDP
Catalog No. 502943427
Encompass_Preferred
A Cdk2 inhibitor (IC50 = 0.044 µM); selective for Cdk2 over Cdk1, Cdk4, Cdk7, and Cdk9 at 100 µM
Catalog No. 50-294-3427 Supplier Cayman Chemical Supplier No. 423355 MG
A Cdk2 inhibitor (IC50 = 0.044 µM); selective for Cdk2 over Cdk1, Cdk4, Cdk7, and Cdk9 at 100 µM