Learn More
Cayman Chemical Cdk2 InhIbItr II 1mg
SDP
Catalog No. 502970904
Encompass_Preferred
A 3-(benzylidne)indolin-2-one analog that selectively and potently inhibits Cdk2 (IC50 = 60 nM)
Catalog No. 50-297-0904 Supplier Cayman Chemical Supplier No. 151541 MG
A 3-(benzylidne)indolin-2-one analog that selectively and potently inhibits Cdk2 (IC50 = 60 nM)