Learn More
Cayman Chemical Cdk2 Inhibitor II, Purity: ≥95%, CAS: 222035-13-4, 5 mg
SDP
Catalog No. 502971607
Encompass_Preferred
A 3-(benzylidne)indolin-2-one analog that selectively and potently inhibits Cdk2 (IC50 = 60 nM)
Catalog No. 50-297-1607 Supplier Cayman Chemical Supplier No. 151545 MG
A 3-(benzylidne)indolin-2-one analog that selectively and potently inhibits Cdk2 (IC50 = 60 nM)