Learn More
Aviva Systems Biology Corp CDK20 Antibody - N-terminal region (ARP62167_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62167_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDK20. It was validated on Western Blot. . Alternate names: CCRK, CDCH, P42, PNQALREApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-594