Learn More
Aviva Systems Biology Corp CDK2AP1 Antibody - C-terminal region (ARP64230_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64230_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDK2AP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human CDK2AP1. Alternate names: DOC1, DORC1, ST19, doc-1, p12DOC-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-208