Learn More
Sino Biological Recombinant Human CDK13/CDC2L5 Protein (GST Tag)
Catalog No. 502575637
Encompass_Preferred
Recombinant human CDK13 (516-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5637 Supplier Sino Biological Supplier No. C2535G100
Recombinant human CDK13 (516-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.